2,6-Difluoro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18974-25G 18974-5G
76897-412EA 418 USD
76897-412 76897-414
2,6-Difluoro-3-methoxybenzaldehyde ≥97%
2,6-Difluoro-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04115899
CAS Number: 149949-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR