2,6-Difluoro-3-isopropoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2,6-Difluoro-3-isopropoxyphenyl)boronic acid

18970-25G 18970-5G
76897-400EA 418 USD
76897-400 76897-402
2,6-Difluoro-3-isopropoxyphenylboronic acid ≥95%
2,6-Difluoro-3-isopropoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BF₂O₃
MW: 215.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07369753
CAS Number: 849062-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR