3-(Dimethylcarbamoyl)-2-fluorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-3-(N,N-dimethylaminocarbonyl)phenylboronic acid, 3-(Dimethylcarbamoyl)-2-Fluorobenzeneboronic Acid

18883-1G 18883-25G 18883-500MG 18883-5G
76897-376EA 290.69 USD
76897-376 76897-378 76897-380 76897-382
3-(Dimethylcarbamoyl)-2-fluorophenylboronic acid ≥95%
3-(Dimethylcarbamoyl)-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BFNO₃
MW: 210.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16295256
CAS Number: 1451391-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR