2,4-Dimethyl-3-fluorobromobenzene ≥97%

Supplier: AOBChem USA

Warning

18872-1G 18872-25G 18872-500MG 18872-5G
76897-362EA 147.47 USD
76897-362 76897-364 76897-366 76897-368
2,4-Dimethyl-3-fluorobromobenzene ≥97%
2,4-Dimethyl-3-fluorobromobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrF
Storage Temperature: Refrigerator
MDL Number: MFCD00079714
CAS Number: 26584-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR