2-Chloro-4-methylnicotinic acid ≥97% research grade

Supplier: AOBChem USA

Warning

18848-1G 18848-25G 18848-500MG 18848-5G
76897-346EA 144.29 USD
76897-346 76897-348 76897-350 76897-352
2-Chloro-4-methylnicotinic acid ≥97% research grade
2-Chloro-4-methylnicotinic acid
Formula: C₇H₆ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11501265
CAS Number: 142266-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR