2-Fluoro-6-(trifluoromethyl)pyridine-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Fluoro-6-(trifluoromethyl)pyridin-3-yl)boronic acid

18840-1G 18840-25G 18840-500MG 18840-5G
76897-334EA 274.78 USD
76897-334 76897-336 76897-338 76897-340
2-Fluoro-6-(trifluoromethyl)pyridine-3-boronic acid ≥97%
2-Fluoro-6-(trifluoromethyl)pyridine-3-boronic acid
Formula: C₆H₄BF₄NO₂
MW: 208.906 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025974
CAS Number: 1150114-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR