4-Cyano-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-4-formylbenzonitrile, 4-Cyano-3-fluorobenzaldehyde

18822-25G 18822-5G
76897-326EA 524.09 USD
76897-326 76897-328
4-Cyano-3-fluorobenzaldehyde ≥97%
4-Cyano-3-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄FNO
MW: 149.1200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738989
CAS Number: 101048-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR