3-Iodopyridine-2-carbonitrile ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Iodopicolinonitrile, 2-Cyano-3-iodopyridine

18742-25G 18742-5G
76897-316EA 524.09 USD
76897-316 76897-318
3-Iodopyridine-2-carbonitrile ≥97%
3-Iodopyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₃IN₂
MW: 230.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09037451
CAS Number: 827616-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR