2-Chloro-6-(trifluoromethyl)benzoyl chloride ≥97% research grade

Supplier: AOBChem USA

Warning

18727-25G 18727-5G
76897-308EA 183.54 USD
76897-308 76897-310
2-Chloro-6-(trifluoromethyl)benzoyl chloride ≥97% research grade
2-Chloro-6-(trifluoromethyl)benzoyl chloride
Formula: C₈H₃Cl₂F₃O
MW: 243.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09025361
CAS Number: 916420-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR