2-Fluoro-3-carboxypyridine-5-boronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

18708-1G 18708-25G 18708-500MG 18708-5G
76897-292EA 201.58 USD
76897-292 76897-294 76897-296 76897-298
2-Fluoro-3-carboxypyridine-5-boronic acid ≥95% research grade
2-Fluoro-3-carboxypyridine-5-boronic acid
Formula: C₆H₅BFNO₄
MW: 184.918 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181650
CAS Number: 1451393-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR