2,6-Difluoropyridine-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Difluoroisonicotinic acid

18656-1G 18656-25G 18656-5G
76897-278EA 62.6 USD
76897-278 76897-280 76897-282
2,6-Difluoropyridine-4-carboxylic acid ≥97%
2,6-Difluoropyridine-4-carboxylic acid
Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09864665
CAS Number: 88912-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR