2,6-Difluoro-3-hydroxypyridine ≥97%

Supplier: AOBChem USA

Warning

18650-1G 18650-25G 18650-5G
76897-268EA 76.39 USD
76897-268 76897-270 76897-272
2,6-Difluoro-3-hydroxypyridine ≥97%
2,6-Difluoro-3-hydroxypyridine
Formula: C₅H₃F₂NO
Storage Temperature: Ambient
MDL Number: MFCD11044351
CAS Number: 209328-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR