2-Fluoro-6-methyl-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-6-methylpyridine-3-boronic acid, 2-Fluoro-6-methylpyridin-3-ylboronic acid

18556-1G 18556-25G 18556-5G
76897-244EA 112.46 USD
76897-244 76897-246 76897-248
2-Fluoro-6-methyl-3-pyridineboronic acid ≥97%
2-Fluoro-6-methyl-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₂
MW: 154.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411560
CAS Number: 906744-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR