2-Bromo-4-methoxy-6-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18522-1G 18522-25G 18522-5G
76897-222EA 83.82 USD
76897-222 76897-224 76897-226
2-Bromo-4-methoxy-6-fluorophenylboronic acid ≥97%
2-Bromo-4-methoxy-6-fluorophenylboronic acid
Formula: C₇H₇BBrFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD12026714
CAS Number: 1315476-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR