(2-Chloro-5-(trifluoromethyl)pyridin-4-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-5-(trifluoromethyl)pyridine-4-boronic acid

18459-1G 18459-25G 18459-500MG 18459-5G
76897-196EA 242.95 USD
76897-196 76897-198 76897-200 76897-202
(2-Chloro-5-(trifluoromethyl)pyridin-4-yl)boronic acid ≥95%
(2-Chloro-5-(trifluoromethyl)pyridin-4-yl)boronic acid
Formula: C₆H₄BClF₃NO₂
MW: 225.360 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12964588
CAS Number: 1167437-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR