2-Ethoxyisonicotinic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Ethoxypyridine-4-carboxylic acid

18419-25G 18419-5G
76897-188EA 375.56 USD
76897-188 76897-190
2-Ethoxyisonicotinic acid ≥95%
2-Ethoxyisonicotinic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.1620 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115719
CAS Number: 91940-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR