2-Chloro-4-ethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18360-1G 18360-25G 18360-500MG 18360-5G
76897-178EA 227.04 USD
76897-178 76897-180 76897-182 76897-184
2-Chloro-4-ethoxybenzaldehyde ≥97%
2-Chloro-4-ethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD23132246
CAS Number: 245368-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR