2-Isopropoxy-5-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (6-Isopropoxypyridin-3-yl)boronic acid, 2-Isoproxypyridine-5-boronic acid

18177-25G 18177-5G
76897-134EA 566.53 USD
76897-134 76897-136
2-Isopropoxy-5-pyridineboronic acid ≥97%
2-Isopropoxy-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₃
Storage Temperature: Ambient
MDL Number: MFCD07368870
CAS Number: 870521-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR