2-Ethoxy-3-methylpyridine-5-boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(6-Ethoxy-5-methylpyridin-3-yl)boronic acid
17987-1G
17987-25G
17987-500MG
17987-5G
76897-072EA
120.95
USD
76897-072
76897-074
76897-076
76897-078
2-Ethoxy-3-methylpyridine-5-boronic acid ≥97%
2-Ethoxy-3-methylpyridine-5-boronic acid
Precaución: for Research Use Only
Formula:
C₈H₁₂BNO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD13181608 CAS Number: 1451391-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...