2-Acetylamino-5-bromo-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

17956-25G 17956-5G
76897-062EA 375.56 USD
76897-062 76897-064
2-Acetylamino-5-bromo-4-methylpyridine ≥97%
2-Acetylamino-5-bromo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₈H₉BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD06798092
CAS Number: 142404-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR