2-Methoxypyrimidine-5-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

17918-25G 17918-5G
76897-050EA 289.63 USD
76897-050 76897-052
2-Methoxypyrimidine-5-carbaldehyde ≥97%
2-Methoxypyrimidine-5-carbaldehyde
Formula: C₆H₆N₂O₂
MW: 138.1200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03446732
CAS Number: 90905-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR