2-Fluoro-5-methylpyridine-4-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (2-Fluoro-5-methylpyridin-4-yl)boronic acid

17898-1G 17898-25G 17898-500MG 17898-5G
76897-036EA 379.81 USD
76897-036 76897-038 76897-040 76897-042
2-Fluoro-5-methylpyridine-4-boronic acid ≥97% research grade
2-Fluoro-5-methylpyridine-4-boronic acid
Formula: C₆H₇BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD11045052
CAS Number: 929194-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR