2-Bromo-3-hydroxy-6-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-5-hydroxy-2-picoline, 2-Bromo-6-methylpyridin-3-ol

17762-25G 17762-5G
76896-992EA 325.7 USD
76896-992 76896-994
2-Bromo-3-hydroxy-6-methylpyridine ≥97%
2-Bromo-3-hydroxy-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06655956
CAS Number: 23003-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR