2-Bromo-6-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

17753-1G 17753-25G 17753-5G
76896-978EA 106.09 USD
76896-978 76896-980 76896-982
2-Bromo-6-(trifluoromethyl)phenol ≥97%
2-Bromo-6-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O
MW: 241.005 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194490
CAS Number: 2844-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR