2-(2-Isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

17745-25G 17745-5G
76896-972EA 251.44 USD
76896-972 76896-974
2-(2-Isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2-Isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₅H₂₃BO₂
Storage Temperature: Ambient
MDL Number: MFCD10698490
CAS Number: 852110-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR