2-Methoxy-4-methyl-5-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Methoxy-4-methylpyridin-3-ylboronic acid, 2-Methoxy-4-methylpyridine-5-boronic acid

17720-1G 17720-25G 17720-5G
76896-954EA 62.6 USD
76896-954 76896-956 76896-958
2-Methoxy-4-methyl-5-pyridineboronic acid ≥97%
2-Methoxy-4-methyl-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₃
Storage Temperature: Ambient
MDL Number: MFCD07368880
CAS Number: 503184-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR