2-Fluoro-4-(piperidine-1-carbonyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

17688-1G 17688-25G 17688-500MG 17688-5G
76896-938EA 171.87 USD
76896-938 76896-940 76896-942 76896-944
2-Fluoro-4-(piperidine-1-carbonyl)phenylboronic acid ≥95%
2-Fluoro-4-(piperidine-1-carbonyl)phenylboronic acid
Formula: C₁₂H₁₅BFNO₃
MW: 251.062 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11856045
CAS Number: 874289-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR