2-[3-Bromo-5-(trifluoromethoxy)phenyl]-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

17662-1G 17662-25G 17662-5G 17662-500MG
76896-914EA 112.46 USD
76896-914 76896-916 76896-918 77644-968
2-[3-Bromo-5-(trifluoromethoxy)phenyl]-1,3-dioxolane ≥95%
2-[3-Bromo-5-(trifluoromethoxy)phenyl]-1,3-dioxolane
Formula: C₁₀H₈BrF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD31619390
CAS Number: 1610379-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR