2,6-Dibromobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

17545-25G 17545-5G
76896-886EA 244.01 USD
76896-886 76896-888
2,6-Dibromobenzoic acid ≥97%
2,6-Dibromobenzoic acid
Formula: C₇H₄Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00234254
CAS Number: 601-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR