2,4-Diethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17530-25G 17530-5G
76896-876EA 354.35 USD
76896-876 76896-878
2,4-Diethoxyphenylboronic acid ≥97%
2,4-Diethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₄
Storage Temperature: Ambient
MDL Number: MFCD09265143
CAS Number: 1072952-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR