2,4-Difluoro-3-formylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
17460-1G
17460-500MG
17460-5G
17460-25G
76896-858EA
558.04
USD
76896-858
76896-860
76896-862
77635-486
2,4-Difluoro-3-formylbenzoic acid ≥97%
2,4-Difluoro-3-formylbenzoic acid
Formula:
C₈H₄F₂O₃ MW: 186.112 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20660537 CAS Number: 1203662-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...