2-Chloro-6-(trifluoromethyl)isonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-6-(trifluoromethyl)pyridine-4-carboxylic acid

17437-1G 17437-25G 17437-5G
76896-850EA 102.91 USD
76896-850 76896-852 76896-854
2-Chloro-6-(trifluoromethyl)isonicotinic acid ≥97%
2-Chloro-6-(trifluoromethyl)isonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO₂
MW: 225.552 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13188811
CAS Number: 796090-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR