2-Bromo-5-chloro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

17414-1G 17414-25G 17414-5G
76896-836EA 74.27 USD
76896-836 76896-838 76896-840
2-Bromo-5-chloro-4-iodopyridine ≥97%
2-Bromo-5-chloro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClIN
MW: 318.338 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16295248
CAS Number: 1061357-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR