3-(Ethoxycarbonyl)-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Ethoxycarbonyl-2-fluorobenzeneboronic acid, 2-Fluoro-3-(ethoxycarbonyl)phenylboronic acid, (3-(Ethoxycarbonyl)-2-fluorophenyl)boronic acid

17390-1G 17390-25G 17390-5G
76896-826EA 70.02 USD
76896-826 76896-828 76896-830
3-(Ethoxycarbonyl)-2-fluorophenylboronic acid ≥97%
3-(Ethoxycarbonyl)-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BFO₄
Storage Temperature: Ambient
MDL Number: MFCD08064050
CAS Number: 1072952-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR