2,4-Difluoro-2-(phenylmethoxy)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

17255-1G 17255-25G 17255-5G 17255-500MG
76896-788EA 84.88 USD
76896-788 76896-790 76896-792 77648-966
2,4-Difluoro-2-(phenylmethoxy)phenylboronic acid ≥97% research grade
2,4-Difluoro-2-(phenylmethoxy)phenylboronic acid
Formula: C₁₃H₁₁BF₂O₃
Storage Temperature: Refrigerator
CAS Number: 3023270-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR