2-Carboxythiophene-4-boronic acid ≥97%

Supplier: AOBChem USA

Warning

17237-1G 17237-25G 17237-500MG 17237-5G
76896-766EA 144.29 USD
76896-766 76896-768 76896-770 76896-772
2-Carboxythiophene-4-boronic acid ≥97%
2-Carboxythiophene-4-boronic acid
Formula: C₅H₅BO₄S
Storage Temperature: Ambient
MDL Number: MFCD08689521
CAS Number: 913835-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR