2',4'-Difluoro-3'-methoxyacetophenone ≥97%
Supplier: AOBChem USA
|
Warning
|
17214-25G
17214-5G
76896-756EA
595.17
USD
76896-756
76896-758
2',4'-Difluoro-3'-methoxyacetophenone ≥97%
2',4'-Difluoro-3'-methoxyacetophenone
Precaución: for Research Use Only
Formula:
C₉H₈F₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD02258906 CAS Number: 373603-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...