6-Bromo-o-anisaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-6-methoxybenzaldehyde

17186-1G 17186-25G 17186-5G
76896-742EA 64.72 USD
76896-742 76896-744 76896-746
6-Bromo-o-anisaldehyde ≥97%
6-Bromo-o-anisaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08059232
CAS Number: 126712-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR