2-Bromo-5-iodo-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

17068-1G 17068-25G 17068-500MG 17068-5G
76896-690EA 379.81 USD
76896-690 76896-692 76896-694 76896-696
2-Bromo-5-iodo-4-methylbenzoic acid ≥97%
2-Bromo-5-iodo-4-methylbenzoic acid
Formula: C₈H₆BrIO₂
MW: 340.941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28130046
CAS Number: 1022983-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR