(2-Bromo-5-iodophenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-3-iodophenylboronic acid

17063-1G 17063-25G 17063-5G
76896-678EA 96.55 USD
76896-678 76896-680 76896-682
(2-Bromo-5-iodophenyl)boronic acid ≥97% research grade
(2-Bromo-5-iodophenyl)boronic acid
Formula: C₆H₅BBrIO₂
Storage Temperature: Refrigerator
MDL Number: MFCD27923891
CAS Number: 1451393-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR