2-Fluoro-3-methylanisole ≥97%

Supplier: AOBChem USA

Warning

16984-1G 16984-25G 16984-5G
76896-670EA 62.6 USD
76896-670 76896-672 76896-674
2-Fluoro-3-methylanisole ≥97%
2-Fluoro-3-methylanisole
Formula: C₈H₉FO
Storage Temperature: Ambient
MDL Number: MFCD09878418
CAS Number: 951885-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR