2-Iodophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16965-1G 16965-25G 16965-5G
76896-662EA 88.06 USD
76896-662 76896-664 76896-666
2-Iodophenylboronic acid ≥97%
2-Iodophenylboronic acid
Formula: C₆H₆BIO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09743934
CAS Number: 1008106-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR