2-(Benzyloxy)-1-bromo-4-fluorobenzene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Benzyloxy-1-bromo-4-fluorobenzene

16942-1G 16942-25G 16942-5G
76896-652EA 93.36 USD
76896-652 76896-654 76896-656
2-(Benzyloxy)-1-bromo-4-fluorobenzene ≥97% research grade
2-(Benzyloxy)-1-bromo-4-fluorobenzene
Formula: C₁₃H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD09033866
CAS Number: 202857-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR