2-Fluoro-3-formyl-4-(trifluoromethyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

16920-1G 16920-25G 16920-500MG 16920-5G
76896-642EA 196.27 USD
76896-642 76896-644 76896-646 76896-648
2-Fluoro-3-formyl-4-(trifluoromethyl)phenylboronic acid ≥97% research grade
2-Fluoro-3-formyl-4-(trifluoromethyl)phenylboronic acid
Formula: C₈H₅BF₄O₃
MW: 235.928 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11856030
CAS Number: 1451393-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR