2-Ethyl-6-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

16847-1G 16847-25G 16847-500MG 16847-5G
76896-618EA 173.99 USD
76896-618 76896-620 76896-622 76896-624
2-Ethyl-6-methoxypyridine ≥95%
2-Ethyl-6-methoxypyridine
Formula: C₈H₁₁NO
MW: 137.179 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15144543
CAS Number: 199273-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR