2-Fluoro-6-methoxy-3-methylphenylboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: (2-Fluoro-6-methoxy-3-methylphenyl)boronic acid

16809-25G 16809-5G
76896-606EA 608.96 USD
76896-606 76896-608
2-Fluoro-6-methoxy-3-methylphenylboronic acid ≥95% research grade
2-Fluoro-6-methoxy-3-methylphenylboronic acid
Formula: C₈H₁₀BFO₃
Storage Temperature: Ambient
MDL Number: MFCD11856029
CAS Number: 1451392-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR