2-Methoxy-4-(trifluoromethoxy)bromobenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1-Bromo-2-methoxy-4-(trifluoromethoxy)benzene
16765-25G
16765-5G
76896-578EA
580.32
USD
76896-578
76896-580
2-Methoxy-4-(trifluoromethoxy)bromobenzene ≥97%
2-Methoxy-4-(trifluoromethoxy)bromobenzene
Precaución: for Research Use Only
Formula:
C₈H₆BrF₃O₂ MW: 271.031 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08458014 CAS Number: 848360-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...