2-Bromo-4-(trifluoromethoxy)benzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Bromo-4-(trifluoromethoxy)phenyl)boronic acid

16762-1G 16762-25G 16762-500MG 16762-5G
76896-568EA 167.63 USD
76896-568 76896-570 76896-572 76896-574
2-Bromo-4-(trifluoromethoxy)benzeneboronic acid ≥97%
2-Bromo-4-(trifluoromethoxy)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BBrF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD19237163
CAS Number: 959997-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR