(2-Methoxy-5-methylpyridin-3-yl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-5-methylpyridine-3-boronic acid

16706-1G 16706-25G 16706-500MG 16706-5G
76896-520EA 287.51 USD
76896-520 76896-522 76896-524 76896-526
(2-Methoxy-5-methylpyridin-3-yl)boronic acid ≥97% research grade
(2-Methoxy-5-methylpyridin-3-yl)boronic acid
Formula: C₇H₁₀BNO₃
Storage Temperature: Ambient
MDL Number: MFCD07437974
CAS Number: 1029654-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR