2-Chloro-5-fluorophenetole ≥95%

Supplier: AOBChem USA

Warning

16490-1G 16490-25G 16490-500MG 16490-5G
76896-470EA 147.47 USD
76896-470 76896-472 76896-474 76896-476
2-Chloro-5-fluorophenetole ≥95%
2-Chloro-5-fluorophenetole
Formula: C₈H₈ClFO
Storage Temperature: Refrigerator
MDL Number: MFCD00672981
CAS Number: 289039-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR